HEPES Buffered Saline Solution 1 * 250 mL
Catalog # C-40020
Supplier: PROMOCELL
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microscope Slides
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Trypsin neutralizing solution
Supplier: PROMOCELL
Trypsin / EDTA (0.04% / 0.03%) 1 * 125 mL
Supplier: PROMOCELL