Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Hand held scrapers

Hand held scrapers

Scraper, white, 246 mm, 112 mm
Catalog # 129-0609
Supplier:   MICRONOVA
Hand held scrapers
Catalog # 129-0609
Supplier:  MICRONOVA
Hand held scrapers

Scraper, white, 246 mm, 112 mm

Price
£37.30
Log in for availability
1 * 1 items
Product Details & Documents

This hand held scraper tool is made with a seamless, one piece injection-mould that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.

  • Injection-moulded, seamless construction
  • Will not scratch metal surfaces
  • Made from FDA approved polypropylene
  • Suitable for indirect food contact
  • Handle shaped for strong grip
  • Autoclavable

The scraper is chemical-resistant and will not rust or corrode.

Specifications
  • Wiping area app.:
    112 mm
  • Description:
    Scraper, white
  • Length:
    246 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.