Fluorescent inks, Blak-Ray®
Ink, A946, UV fluorescing non-porous, normal color/colorless, fluorescent color/blue, Industrial coding, security print, coupons/tickets, inventory control
Catalog # UVPA96-0057-04
Supplier: UVP ULTRA VIOLET PRODUCTS
Jump to:
Product Details & Documents
Blak-Ray® fluorescent inks are suited for use on skin, fabric and similar porous or textured surfaces.
- Simple to use - inks come ready made
- Won't gum up - stamp pads stay cleaner longer
- Economical - small amounts of ink are used with stamp pads
- All inks are formulated to fluoresce brightly under UVP's longwave ultraviolet lamps
- Readmission type inks are designed for use at amusement parks, social gatherings and sporting events
- Inks are non-toxic, fast-drying and non-transferable
Suitable for readmission control, inspection, coding and inventory control.
Specifications
- For Use With:Industrial coding, security print, coupons/tickets, inventory control
- Description:Ink, A946, UV fluorescing non-porous, normal color/colorless, fluorescent color/blue
- Seq:03