Colony counters, ProtoCOL 3
Automated colony counter with zone sizing and validation plates, 3
Catalog # SYNBPROC3
Supplier: SYNBIOSIS
Jump to:
Product Details & Documents
ProtoCOL 3 is designed for colony counting, zone measuring (inhibition and AST), membranes, Petri-film™ and other applications including spiral plates, OPKA, SBA, Multi-sector, Multi-well, SRD and Ames.
- High throughput capability suitable for production and large scale testing environments
- Extensive report generation to Excel or a LIMS system
- Separation of touching colonies, exclusion areas, colour mode, shape mode, size mode
- High resolution colour images ensure that the smallest colony can be seen and counted with zone measurements
- Mounted touch screen processor, which enables ease of control while the easy to use software produces highly accurate results
Designed for use across a wide range of applications, ProtoCOL 3 will count colonies as small as 43 µm (0,043 mm) or measure zones accurately to 0,1 mm.
Certifications: GLP/GMP compliant data.
Specifications
- Model:3
- Description:Automated colony counter with zone sizing and validation plates
- Cat. No.:SYNBPROC3
- Dish size:150 mm
- W×D×H:400×500×450 mm
- Illumination:Two sliding doors to prevent excessive ambient light
Unique three channels: Red, green, blue
LED lighting (patent pending)
Multi-array computer controlled LED
Lower lighting with upper reflective lighting for all applications - Camera:Scientific grade CCD
- Weight:20 kg
- Resolution:1,4 MP
Accessories
Product Information
- Catalog #AvailabilityPrice
- Scanner with software
Product Information
- Catalog #AvailabilityPrice
- SBA software module for serum bactericidal assaySBA software module for serum bactericidal assaySYNBPROC3-SBA-MODLog in for availability1 * 1 items
Product Information
- Catalog #AvailabilityPrice
- Counting software module for spiral plates, includes validation plateCounting software module for spiral plates, includes validation plateSYNBPROC3SPIRALMODLog in for availability1 * 1 items - £1,330.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



