INVIVOGEN OX-PAPC 1 * 1 mg
Catalog # INVVTLRL-OXP1
Supplier: Invivogen
Frequently Bought Together
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERILIN
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose
Supplier: Cytiva
Centrifuge tubes, SuperClear®
Supplier: LABCON
Sterilised write-on bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cremophor EL®, Sterilized 1 * 1 SET
Supplier: Merck Millipore (Calbiochem)
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Calibrated Disposable Plastic Inoculation Loops
Supplier: Copan
DYNGO-4A 1 * 10 mg
Supplier: Cayman Chemical
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Detectable scoops, SteriPlast®
Supplier: BURKLE
Reagent/centrifuge tubes, flat base, with screw cap
Supplier: SARSTEDT
Sampling spoons, eco-friendly, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP