Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Hand held scrapers

Hand held scrapers

Scraper, white, 203 mm, 76 mm
  129-0608
 :   MICRONOVA
Hand held scrapers
  129-0608
 :  MICRONOVA
Hand held scrapers

Scraper, white, 203 mm, 76 mm

 
£31.50
   
1 * 1 items
 

This hand held scraper tool is made with a seamless, one piece injection-mould that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.

  • Injection-moulded, seamless construction
  • Will not scratch metal surfaces
  • Made from FDA approved polypropylene
  • Suitable for indirect food contact
  • Handle shaped for strong grip
  • Autoclavable

The scraper is chemical-resistant and will not rust or corrode.

 
  • Wiping area app.:
    76 mm
  • Description:
    Scraper, white
  • Length:
    203 mm
  • Seq:
    01