Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Microvolume inserts, CrossLab

Microvolume inserts, CrossLab

Flat bottom, for 8 mm screw top vials, 0,20 ml, Glass, No
Catalog # HEWL8010-0126
Supplier:   Agilent
Microvolume inserts, CrossLab
Catalog # HEWL8010-0126
Supplier:  Agilent
Microvolume inserts, CrossLab

Flat bottom, for 8 mm screw top vials, 0,20 ml, Glass, No

Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
Specifications
  • Cap included:
    No
  • Capacity:
    0,20 ml
  • Description:
    Flat bottom, for 8 mm screw top vials
  • Material:
    Glass
  • Seq:
    03