FOLINIC ACID CALCIUM SALT HYDRATE BIOXTRA, >= 99.0% (HPLC) 1 * 1 g
Catalog # SIAL47612-1G
Supplier: Merck
FOLINIC ACID CALCIUM SALT HYDRATE BIOXTRA, >= 99.0% (HPLC) 1 * 1 g
Catalog # SIAL47612-1G
Supplier: Merck
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals