ω-Bromo-4-nitroacetophenone
Glass bottle for solids, 25 g
Catalog # FLUO018905-25G
Marketsource
Supplier: FLUOROCHEM
ω-Bromo-4-nitroacetophenone
Catalog # FLUO018905-25G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 99-81-0
Jump to:
Product Details & Documents
Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
- CAS Number:99-81-0
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00007356
- Molecular Weight:244.04 g/mol
- Melting Pt:100 °C
- Storage Temperature:Ambient
- EINECS:202-789-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



