ω-Bromo-4-nitroacetophenone
Glass bottle for solids, 100 g
Catalog # FLUO018905-100G
Marketsource
Supplier: FLUOROCHEM
ω-Bromo-4-nitroacetophenone
Catalog # FLUO018905-100G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 99-81-0
Jump to:
Product Details & Documents
Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
- CAS Number:99-81-0
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00007356
- Molecular Weight:244.04 g/mol
- Melting Pt:100 °C
- Storage Temperature:Ambient
- EINECS:202-789-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



