2-Ethoxybenzonitrile
Glass bottle for solids, 25 g
Catalog # FLUO018661-25G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:6609-57-0
- Formula:C₉H₉NO
- MDL Number:MFCD00001789
- Molecular Weight:147.18 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



