Order Entry
United Kingdom
ContactUsLinkComponent
Coated GC Packing, phase 4% Carbowax 20M + 0.8% KOH, matrix 60/80 Carbopack B support, bottle of 15 g 1 * 15 g
Catalog # 11887.
Supplier:  Merck
Coated GC Packing, phase 4% Carbowax 20M + 0.8% KOH, matrix 60/80 Carbopack B support, bottle of 15 g 1 * 15 g
Catalog # 11887.
Supplier:  Merck
Frequently Bought Together