Order Entry
United Kingdom
ContactUsLinkComponent
5-Bromo-2-(4-BOC-piperazin-1-yl)-3-methylpyridine , ECN [878809-70-2], formula C15H22BrN3O2, storage room tempratre, transport norma l temperature, FW 356.3 1 * 1 g
Catalog # COBBOT-8026-1G
Supplier:  COMBI-BLOCKS
5-Bromo-2-(4-BOC-piperazin-1-yl)-3-methylpyridine , ECN [878809-70-2], formula C15H22BrN3O2, storage room tempratre, transport norma l temperature, FW 356.3 1 * 1 g
Catalog # COBBOT-8026-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together