Order Entry
United Kingdom
ContactUsLinkComponent
5-Iodo-1,3-dimethylpyrimidine-2,4-dione , ECN [40738-83-8], formula C6H7IN2O2, storage room tempratre, transport normal temperature, FW 266.0 1 * 1 g
Catalog # COBBPY-1392-1G
Supplier:  COMBI-BLOCKS
5-Iodo-1,3-dimethylpyrimidine-2,4-dione , ECN [40738-83-8], formula C6H7IN2O2, storage room tempratre, transport normal temperature, FW 266.0 1 * 1 g
Catalog # COBBPY-1392-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together