3-Fluoropyrocatechol
Glass bottle for solids, 100 g
Catalog # COBBOS-7647-100G
Marketsource
Supplier: COMBI-BLOCKS
3-Fluoropyrocatechol
Catalog # COBBOS-7647-100G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 363-52-0
Jump to:
Product Details & Documents
Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
- CAS Number:363-52-0
- Formula:C₆H₅FO₂
- MDL Number:MFCD00042582
- Molecular Weight:128.1 g/mol
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:71 °C
- Density:1,514 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



