Order Entry
United Kingdom
ContactUsLinkComponent
3-Bromo-1-isopropyl-5-(trifluoromethyl)pyridin-2(1H)-one, ECN [1215205-40-5], formula C9H9BrF3NO, storage room tempratre, transport normal temperature, FW 284.1 1 * 5 g
Catalog # COBBPY-1300-5G
Supplier:  COMBI-BLOCKS
3-Bromo-1-isopropyl-5-(trifluoromethyl)pyridin-2(1H)-one, ECN [1215205-40-5], formula C9H9BrF3NO, storage room tempratre, transport normal temperature, FW 284.1 1 * 5 g
Catalog # COBBPY-1300-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together