Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-1-butyl-1H-pyrazole , ECN [957062-61-2], formula C7H11BrN2, storage room tempratre, transport normal temperature, FW 203.1 1 * 1 g
Catalog # COBBHC-6328-1G
Supplier:  COMBI-BLOCKS
4-Bromo-1-butyl-1H-pyrazole , ECN [957062-61-2], formula C7H11BrN2, storage room tempratre, transport normal temperature, FW 203.1 1 * 1 g
Catalog # COBBHC-6328-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together