Order Entry
United Kingdom
ContactUsLinkComponent
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine, ECN [114833-95-3], formula C7H5ClN4, storage room tempratre, transport normal temperature, F W 180.6 1 * 5 g
Catalog # COBBHC-6098-5G
Supplier:  COMBI-BLOCKS
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine, ECN [114833-95-3], formula C7H5ClN4, storage room tempratre, transport normal temperature, F W 180.6 1 * 5 g
Catalog # COBBHC-6098-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together