3-(4-Chlorophenyl)propionic acid
Glass bottle for solids, 5 g
Catalog # COBBHC-2050-5G
Marketsource
Supplier: COMBI-BLOCKS
3-(4-Chlorophenyl)propionic acid
Catalog # COBBHC-2050-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 2019-34-3
Jump to:
Product Details & Documents
Synonyms: 3-(4-Chlorophenyl)propanoic acid
- CAS Number:2019-34-3
- Formula:C₉H₉ClO₂
- MDL Number:MFCD00016555
- Molecular Weight:184.62 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



