Order Entry
United Kingdom
ContactUsLinkComponent
4-(N-Isopropyl-N-(4-methoxybenzyl)sulfamoyl)phenylboronic acid , ECN [913835-96-8], formula C17H22BNO5S, storage room tempratre, tra nsport normal temperature, FW 363.2 1 * 1 g
Catalog # COBBBB-3533-1G
Supplier:  COMBI-BLOCKS
4-(N-Isopropyl-N-(4-methoxybenzyl)sulfamoyl)phenylboronic acid , ECN [913835-96-8], formula C17H22BNO5S, storage room tempratre, tra nsport normal temperature, FW 363.2 1 * 1 g
Catalog # COBBBB-3533-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together