Order Entry
United Kingdom
ContactUsLinkComponent
1-Bromo-4-(tert-butoxymethyl)benzene, ECN [22807-80-3], formula C11H15BrO, storage room tempratre, transport normal temperature, FW 243.1 1 * 5 g
Catalog # COBBOT-1176-5G
Supplier:  COMBI-BLOCKS
1-Bromo-4-(tert-butoxymethyl)benzene, ECN [22807-80-3], formula C11H15BrO, storage room tempratre, transport normal temperature, FW 243.1 1 * 5 g
Catalog # COBBOT-1176-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together