Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

3-Bromophenetole

Glass bottle for solids, 1 g
Catalog # COBBOT-0755-1G
Marketsource
Supplier:   COMBI-BLOCKS
3-Bromophenetole
Catalog # COBBOT-0755-1G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  2655-84-7
3-Bromophenetole

Glass bottle for solids, 1 g

CAS Number:  2655-84-7
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
  • CAS Number:2655-84-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00274287
  • Molecular Weight:201.06 g/mol
  • EINECS:220-186-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications