Order Entry
United Kingdom
ContactUsLinkComponent
4-(3-Chloro-2-fluorophenyl)-2-methylbut-3-yn-2-ol, ECN [1187385-72-3], formula C11H10ClFO, storage room tempratre, transport normal temperature, FW 212.6 1 * 5 g
Catalog # COBBHC-3075-5G
Supplier:  COMBI-BLOCKS
4-(3-Chloro-2-fluorophenyl)-2-methylbut-3-yn-2-ol, ECN [1187385-72-3], formula C11H10ClFO, storage room tempratre, transport normal temperature, FW 212.6 1 * 5 g
Catalog # COBBHC-3075-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together