5-(Hydroxymethyl)indole
Glass bottle for solids, 5 g
Catalog # COBBIN-0009-5G
Marketsource
Supplier: COMBI-BLOCKS
5-(Hydroxymethyl)indole
Catalog # COBBIN-0009-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 1075-25-8
Jump to:
Product Details & Documents
Synonyms: (1H-Indol-5-yl)methanol
- CAS Number:1075-25-8
- Formula:C₉H₉NO
- MDL Number:MFCD02179594
- Molecular Weight:147.1700 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



