Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

4-Fluoroindole

Glass bottle for solids, 1 g
Catalog # COBBIN-0051-1G
Irritant
Marketsource
Supplier:   COMBI-BLOCKS
4-Fluoroindole
Catalog # COBBIN-0051-1G
Supplier:  COMBI-BLOCKS
Marketsource
CAS Number:  387-43-9
4-Fluoroindole

Glass bottle for solids, 1 g

CAS Number:  387-43-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Fluoro-1H-indole
  • CAS Number:387-43-9
  • Formula:C₈H₆FN
  • MDL Number:MFCD00055992
  • Molecular Weight:135.14 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications