Order Entry
United Kingdom
ContactUsLinkComponent
6-Bromo-1-(phenylsulfonyl)-1H-indole , ECN [679794-03-7], formula C14H10BrNO2S, storage room tempratre, transport normal temperature , FW 336.2 1 * 1 g
Catalog # COBBIN-0237-1G
Supplier:  COMBI-BLOCKS
6-Bromo-1-(phenylsulfonyl)-1H-indole , ECN [679794-03-7], formula C14H10BrNO2S, storage room tempratre, transport normal temperature , FW 336.2 1 * 1 g
Catalog # COBBIN-0237-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together