4-Bromo-1-methyl-1H-indole, ECN [590417-55-3], formula C9H8BrN, storage room tempratre, transport normal temperature, FW 210.1 1 * 5 g
Catalog # COBBIN-0221-5G
Supplier: COMBI-BLOCKS
4-Bromo-1-methyl-1H-indole, ECN [590417-55-3], formula C9H8BrN, storage room tempratre, transport normal temperature, FW 210.1 1 * 5 g
Catalog # COBBIN-0221-5G
Supplier: COMBI-BLOCKS
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Glycerine, Hi-LR™
Supplier: HIMEDIA
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
ISE standards
Supplier: Hach
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific