£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
4-Iodo-1-tosyl-1H-imidazole, ECN [163854-63-5], formula C10H9IN2O2S, storage room tempratre, transport normal temperature, FW 348.2 1 * 25 g
COBBHI-1340-25G
:
COMBI-BLOCKS
4-Iodo-1-tosyl-1H-imidazole, ECN [163854-63-5], formula C10H9IN2O2S, storage room tempratre, transport normal temperature, FW 348.2 1 * 25 g
COBBHI-1340-25G
:
COMBI-BLOCKS
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals