Order Entry
United Kingdom
ContactUsLinkComponent
4-Iodo-1-tosyl-1H-imidazole, ECN [163854-63-5], formula C10H9IN2O2S, storage room tempratre, transport normal temperature, FW 348.2 1 * 5 g
Catalog # COBBHI-1340-5G
Supplier:  COMBI-BLOCKS
4-Iodo-1-tosyl-1H-imidazole, ECN [163854-63-5], formula C10H9IN2O2S, storage room tempratre, transport normal temperature, FW 348.2 1 * 5 g
Catalog # COBBHI-1340-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together