Order Entry
United Kingdom
ContactUsLinkComponent
5-Iodo-1-methyl-1H-imidazole , ECN [71759-88-1], formula C4H5IN2, storage room tempratre, transport normal temperature, FW 208.0 1 * 1 g
Catalog # COBBHI-1005-1G
Supplier:  COMBI-BLOCKS
5-Iodo-1-methyl-1H-imidazole , ECN [71759-88-1], formula C4H5IN2, storage room tempratre, transport normal temperature, FW 208.0 1 * 1 g
Catalog # COBBHI-1005-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together