Order Entry
United Kingdom
ContactUsLinkComponent
4-Chloro-6-(4-iodo-1H-pyrazol-1-yl)pyrimidine , ECN [957035-27-7], formula C7H4ClIN4, storage room tempratre, transport normal tempe rature, FW 306.5 1 * 1 g
Catalog # COBBHC-6088-1G
Supplier:  COMBI-BLOCKS
4-Chloro-6-(4-iodo-1H-pyrazol-1-yl)pyrimidine , ECN [957035-27-7], formula C7H4ClIN4, storage room tempratre, transport normal tempe rature, FW 306.5 1 * 1 g
Catalog # COBBHC-6088-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together