Order Entry
United Kingdom
ContactUsLinkComponent
3-(N-t-butyl-N-methylsulfamoyl)phenylboronic acid , ECN [1217501-22-8], formula C11H18BNO4S, storage room tempratre, transport norma l temperature, FW 271.1 1 * 1 g
Catalog # COBBBB-6005-1G
Supplier:  COMBI-BLOCKS
3-(N-t-butyl-N-methylsulfamoyl)phenylboronic acid , ECN [1217501-22-8], formula C11H18BNO4S, storage room tempratre, transport norma l temperature, FW 271.1 1 * 1 g
Catalog # COBBBB-6005-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together