Order Entry
United Kingdom
ContactUsLinkComponent
5-Chloro-2-fluoro-4-methoxyphenylboronic acid , ECN [1072952-18-1], formula C7H7BClFO3, storage room tempratre, transport normal tem perature, FW 204.4 1 * 1 g
Catalog # COBBBB-8465-1G
Supplier:  COMBI-BLOCKS
5-Chloro-2-fluoro-4-methoxyphenylboronic acid , ECN [1072952-18-1], formula C7H7BClFO3, storage room tempratre, transport normal tem perature, FW 204.4 1 * 1 g
Catalog # COBBBB-8465-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together