Methyl 4-bromo-1H-pyrrole-2-carboxylate, ECN [934-05-4], formula C6H6BrNO2, storage room tempratre, transport normal temperature, FW 204.0 1 * 5 g
Catalog # COBBHI-1441-5G
Supplier: COMBI-BLOCKS
Methyl 4-bromo-1H-pyrrole-2-carboxylate, ECN [934-05-4], formula C6H6BrNO2, storage room tempratre, transport normal temperature, FW 204.0 1 * 5 g
Catalog # COBBHI-1441-5G
Supplier: COMBI-BLOCKS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection