5-Bromo-1-tert-butyl-6-fluorobenzoimidazole , ECN [1187385-67-6], formula C11H12BrFN2, storage room tempratre, transport normal temp erature, FW 271.1 1 * 1 g
Catalog # COBBHI-1531-1G
Supplier: COMBI-BLOCKS
5-Bromo-1-tert-butyl-6-fluorobenzoimidazole , ECN [1187385-67-6], formula C11H12BrFN2, storage room tempratre, transport normal temp erature, FW 271.1 1 * 1 g
Catalog # COBBHI-1531-1G
Supplier: COMBI-BLOCKS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection