Order Entry
United Kingdom
ContactUsLinkComponent
1,2-Bis(5-nitropyridin-2-yl)disulfane, ECN [2127-10-8], formula C10H6N4O4S2, storage room tempratre, transport normal temperature, F W 310.3 1 * 5 g
Catalog # COBBPY-7399-5G
Supplier:  COMBI-BLOCKS
1,2-Bis(5-nitropyridin-2-yl)disulfane, ECN [2127-10-8], formula C10H6N4O4S2, storage room tempratre, transport normal temperature, F W 310.3 1 * 5 g
Catalog # COBBPY-7399-5G
Supplier:  COMBI-BLOCKS
Frequently Bought Together