Order Entry
United Kingdom
ContactUsLinkComponent
4-(5-Chloro-3-fluoro-2-pyridinyl)morpholine , ECN [1020253-17-1], formula C9H10ClFN2O, storage room tempratre, transport normal temp erature, FW 216.6 1 * 1 g
Catalog # COBBPY-7297-1G
Supplier:  COMBI-BLOCKS
4-(5-Chloro-3-fluoro-2-pyridinyl)morpholine , ECN [1020253-17-1], formula C9H10ClFN2O, storage room tempratre, transport normal temp erature, FW 216.6 1 * 1 g
Catalog # COBBPY-7297-1G
Supplier:  COMBI-BLOCKS
Frequently Bought Together