2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, ECN [72587-15-6], formula C6H2ClF3N2O2, storage room tempratre, transport normal tempe rature, FW 226.5 1 * 5 g
Catalog # COBBPY-1418-5G
Supplier: COMBI-BLOCKS
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, ECN [72587-15-6], formula C6H2ClF3N2O2, storage room tempratre, transport normal tempe rature, FW 226.5 1 * 5 g
Catalog # COBBPY-1418-5G
Supplier: COMBI-BLOCKS
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals