Methanol
205 L
BARRMETH205
Marketsource
Methanol
BARRMETH205
Marketsource
: 67-56-1
: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- :67-56-1
- :CH₃OH
- :MFCD00004595
- :32.04 g/mol
- :13,05984
- :64.6 °C (1013 hPa)
- :–98 °C
- :0.7918 g/cm³ (20 °C)
- :11 °C (closed cup)
- :Ambient
- :200-659-6
- :1230
- :3,II
- :01-2119433307-44
- Pk:205 L
- Cat. No.:BARRMETH205
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



