Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Objectives for CKX53
Objectives for CKX53
Objective for relief contrast
Catalog # OLYMN1507900
Supplier:   OLYMPUS OPTICAL
Objectives for CKX53
Catalog # OLYMN1507900
Supplier:  OLYMPUS OPTICAL
Objectives for CKX53
Objectives for CKX53
Objective for relief contrast
Price
£1,720.00
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    10×
  • Description:
    Objective for relief contrast