MICROTOME BLADES TYPE S 35 1 * 50 items
Catalog # FEAT207500000_B
Supplier: PFM Medical GmbH
Frequently Bought Together
Dewar vessels, cylindrical, type C and G-C
Supplier: KGW
VWR® UV-1600PC, UV/Visible Spectrophotometer
Supplier: VWR Collection
Storage phosphor screens and cassettes
Supplier: Cytiva
VWR® VisiScope® 260, Stereo Microscopes
Supplier: VWR Collection
Barbed luer adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
CRS ZPure™ Hydrocarbon Gas Filters
Supplier: Trajan Scientific and Medical
ADAPTER BNC SOCKET/DIN PLUG TYPE Z862 1 * 1 items
Supplier: SI Analytics
Short-term tubes for accuro®
Supplier: Dräger
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Conductivity meters, handheld, Seven2Go™ S3 / S7 Pro
Supplier: Mettler - Toledo
Accessories for Liquid Nitrogen Vessels, TR,TP and GT
Supplier: Air Liquide
Accessories for meters 765 / 766
Supplier: KNICK
Gas detection tubes for sulphur dioxide
Supplier: Dräger
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
CRS SPure™ H₂O/HC Filters
Supplier: Trajan Scientific and Medical
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
pH/mV/°C meters, bench, SevenExcellence™ S400
Supplier: Mettler - Toledo