Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Phosphate test, RP for 1.18388.0001, 1.14449.0001, Vanadato-molybdate, MCD, 300 Tests
: Merck
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
1.18466.0002
: Merck
:
Quick tests, visual colour comparison.
- Results in approximately 1 minute
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
- Chemical method:Vanadato-molybdate
- Pk:300 Tests
- Description:Phosphate test, RP for 1.18388.0001, 1.14449.0001
- Type:MCD