Order Entry
United Kingdom
ContactUsLinkComponent
Shaft material Glass Measuring range 10 ... 1'000'000 Measuring unit uS/cm Temp.-long time (°C) 5 ... 70 Temp.-short time. (°C) 5 ... 70 Shaft diameter top (mm) 12 Shaft diameter bottom (mm) 20 Shaft length till head (mm) 125 Minimum immersion depth 1 * 1 items
Catalog # METR6.0901.260
Supplier:  METROHM
Shaft material Glass Measuring range 10 ... 1'000'000 Measuring unit uS/cm Temp.-long time (°C) 5 ... 70 Temp.-short time. (°C) 5 ... 70 Shaft diameter top (mm) 12 Shaft diameter bottom (mm) 20 Shaft length till head (mm) 125 Minimum immersion depth 1 * 1 items
Catalog # METR6.0901.260
Supplier:  METROHM
Frequently Bought Together