4'-Hydroxyacetophenone, Sigma-Aldrich®
Glass bottle for solids, 100 g
Catalog # SIAL278564-100G
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:313 °C (1013 hPa)
- Melting Pt:105…108 °C
- Density:1.139 g/cm³ (25 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:202-802-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



