(±)-1,2-Hexanediol, Sigma-Aldrich®
Glass bottle for solids, 50 g
Catalog # SIAL213691-50G
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
- CAS Number:6920-22-5
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00010737
- Molecular Weight:118.18 g/mol
- Boiling Pt:223 °C (1013 hPa)
- Melting Pt:45 °C
- Density:0.9507 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- EINECS:230-029-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



