Methanol
25 L
BARRMETH025
Marketsource
Methanol
BARRMETH025
Marketsource
: 67-56-1
: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- :67-56-1
- :CH₃OH
- :MFCD00004595
- :32.04 g/mol
- :13,05984
- :64.6 °C (1013 hPa)
- :–98 °C
- :0.7918 g/cm³ (20 °C)
- :11 °C (closed cup)
- :Ambient
- :200-659-6
- :1230
- :3,II
- :01-2119433307-44
- Pk:25 L
- Cat. No.:BARRMETH025
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



