Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Racks for freezer

Racks for freezer

16 (4×4), 100 mm, 566×140×319 mm
Catalog # RATI5400005
Supplier:   RATIOLAB
Racks for freezer
Catalog # RATI5400005
Supplier:  RATIOLAB
Racks for freezer

16 (4×4), 100 mm, 566×140×319 mm

Price
£209.00
Log in for availability
1 * 1 items
Product Details & Documents

For the storing of cryoboxes in freezer cabinets and chests.

  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning

Delivery information: Cryo racks are supplied with shelf sizes for any standard cryoboxes.

Specifications
  • Compartments:
    16 (4×4)
  • W×D×H ext.:
    566×140×319 mm
  • Seq:
    05
  • Cryobox height:
    100 mm