Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Racks for freezer

Racks for freezer

4 (1×4), 50 mm, 140×140×222 mm
Catalog # RATI5400041
Supplier:   RATIOLAB
Racks for freezer
Catalog # RATI5400041
Supplier:  RATIOLAB
Racks for freezer

4 (1×4), 50 mm, 140×140×222 mm

Price
£117.00
Log in for availability
1 * 1 items
Product Details & Documents

For the storing of cryoboxes in freezer cabinets and chests.

  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning

Delivery information: Cryo racks are supplied with shelf sizes for any standard cryoboxes.

Specifications
  • Compartments:
    4 (1×4)
  • W×D×H ext.:
    140×140×222 mm
  • Seq:
    08
  • Cryobox height:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.