Racks for freezer
15 (1×15), 32 mm, 140×140×552 mm
Catalog # RATI5400036
Supplier: RATIOLAB
Jump to:
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
- Robust construction
- Handles for convenient transport
- Made of stainless steel with surfaces for perfect cleaning
: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Specifications
- Compartments:15 (1×15)
- W×D×H ext.:140×140×552 mm
- Seq:14
- Cryobox height:32 mm



