Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Wipers for general applications, PIG®
Wipers for general applications, PIG®
Wipes, multi-purpose, in pop-up box, Blue, 420×240 mm, 75 wipes/box
Catalog # NEWPWIP231
Supplier:   NEW PIG
Wipers for general applications, PIG®
Catalog # NEWPWIP231
Supplier:  NEW PIG
Wipers for general applications, PIG®
Wipers for general applications, PIG®
Wipes, multi-purpose, in pop-up box, Blue, 420×240 mm, 75 wipes/box
Price
£149.00
Log in for availability
1 * 900 items
Product Details & Documents
These cellulose wipes with adhesive latex binder are ideal for all kinds of jobs - from cleaning parts to cleaning hands.
  • Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping
  • Made thick and absorbent to quickly soak up oils, degreasers and other liquids
  • Low-lint material won't leave residue or create a mess
  • Excellent for a variety of wiping applications from quick cleanups to degreasing
  • Available 1/4 folded and poly wrapped, as economical rolls or in convenient pop-up boxes
Specifications
  • Packed:
    75 wipes/box
  • Colour:
    Blue
  • Description:
    Wipes, multi-purpose, in pop-up box
  • L×W:
    420×240 mm
  • Seq:
    05